Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc8c
Entrez Gene ID
100604
Gene Name
leucine rich repeat containing 8 family, member C
Gene Aliases
AD158, AI326115, E430036I04Rik, fad158
Chromosome Location
Chr.5: 105519386 - 105613018 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133897.2 NP_598658.1 2 - 3 237 77
GenBank mRNA AB081508.1 - 2 - 3 237 77
AK142337.1 - 2 - 3 267 77
AK169492.1 - 2 - 3 300 77
AK171313.1 - 2 - 3 319 77
BC026572.1 - 2 - 3 247 77

View Full Product Details