The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133931.4 | NP_598692.1 | 15 - 16 | 1995 | 55 |
GenBank mRNA | AK036052.1 | - | 15 - 16 | 1996 | 55 |
AK040864.1 | - | 15 - 16 | 1988 | 55 | |
BC016121.1 | - | 15 - 16 | 1957 | 55 |