Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Isy1
Entrez Gene ID
57905
Gene Name
ISY1 splicing factor homolog (S. cerevisiae)
Gene Aliases
5830446M03Rik, AI181014, AU020769
Chromosome Location
Chr.6: 87818447 - 87838759 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133934.4 NP_598695.1 8 - 9 653 83
GenBank mRNA AK017996.1 - 8 - 9 570 83
AK173116.1 - - 621 83
BC020103.1 - 8 - 9 617 83
BC022597.1 - 1 - 2 125 83
BC037695.1 - 8 - 9 658 83

View Full Product Details