The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040684.1 | NP_001035774.1 | 2 - 3 | 229 | 70 |
NM_133943.2 | NP_598704.2 | 2 - 3 | 281 | 70 | |
GenBank mRNA | AF277718.1 | - | 2 - 3 | 217 | 70 |
AK077476.1 | - | 2 - 3 | 261 | 70 | |
BB854484.1 | - | 2 - 3 | 288 | 70 | |
BC006822.1 | - | 2 - 3 | 221 | 70 | |
BC012954.1 | - | 1 - 2 | 82 | 70 | |
BC018525.1 | - | 2 - 3 | 221 | 70 | |
BC132605.1 | - | 2 - 3 | 215 | 70 | |
BC138589.1 | - | 2 - 3 | 215 | 70 | |
BY133253.1 | - | 2 - 3 | 193 | 70 |