The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159591.1 | NP_001153063.1 | 7 - 8 | 1082 | 89 |
NM_134002.2 | NP_598763.1 | 6 - 7 | 1259 | 89 | |
GenBank mRNA | AK077076.1 | - | 7 - 8 | 1062 | 89 |
AK132871.1 | - | 7 - 8 | 1083 | 89 | |
AK152413.1 | - | 6 - 7 | 1258 | 89 | |
AK206964.1 | - | 1 - 2 | 73 | 89 | |
BC004839.1 | - | 6 - 7 | 1201 | 89 | |
BC063083.1 | - | 4 - 5 | 2383 | 89 |