The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170430.1 | NP_001163901.1 | 8 - 9 | 1103 | 67 |
NM_001170431.1 | NP_001163902.1 | 8 - 9 | 1238 | 67 | |
NM_134060.4 | NP_598821.1 | 8 - 9 | 1081 | 67 | |
GenBank mRNA | AK076592.1 | - | 8 - 9 | 1076 | 67 |
AK077208.1 | - | 8 - 9 | 1057 | 67 | |
AK141280.1 | - | 8 - 9 | 1239 | 67 | |
BC006881.1 | - | 8 - 9 | 1095 | 67 |