Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tnfaip8
Entrez Gene ID
106869
Gene Name
tumor necrosis factor, alpha-induced protein 8
Gene Aliases
AA987150, E130304C20Rik, ENSMUSG00000073567, Gg2-1, Gm10539, Nded, Ssc-2
Chromosome Location
Chr.18: 49979427 - 50093229 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134131.2 NP_598892.2 1 - 2 339 112
GenBank mRNA AK087478.1 - 1 - 2 257 112
AK090316.1 - 1 - 2 149 112
AK090350.1 - 1 - 2 225 112
AK149901.1 - 1 - 2 115 112
AK150445.1 - 1 - 2 111 112
AK150965.1 - 1 - 2 121 112
AK151622.1 - 1 - 2 111 112
AK152426.1 - 1 - 2 111 112
BC009090.1 - 1 - 2 101 112

View Full Product Details