The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138582.1 | NP_613048.1 | 7 - 8 | 1292 | 77 |
GenBank mRNA | AF134319.1 | - | 7 - 8 | 1292 | 77 |
BC137792.1 | - | 7 - 8 | 1233 | 77 | |
BC144911.1 | - | 7 - 8 | 1233 | 77 |