Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phf21a
Entrez Gene ID
192285
Gene Name
PHD finger protein 21A
Gene Aliases
80kDa, Bhc, Bhc80, D030065N23Rik, PFTF1
Chromosome Location
Chr.2: 92183746 - 92364667 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109690.1 NP_001103160.1 8 - 9 1117 63
NM_001109691.1 NP_001103161.1 11 - 12 1273 63
NM_138755.2 NP_620094.2 10 - 11 1276 63
GenBank mRNA AB105178.1 - 10 - 11 1182 63
AK037174.1 - 11 - 12 1274 63
AK129424.1 - 10 - 11 2531 63
AK137085.1 - 7 - 8 1084 63
BC019181.1 - 10 - 11 1131 63

View Full Product Details