Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atxn7
Entrez Gene ID
246103
Gene Name
ataxin 7
Gene Aliases
A430107N12Rik, AI627028, Sca7, ataxin-7
Chromosome Location
Chr.14: 13961439 - 14107302 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139227.4 NP_631973.3 3 - 4 631 99
GenBank mRNA AF455111.1 - 2 - 3 371 99
AK046679.1 - 3 - 4 632 99
BC035459.1 - 2 - 3 284 99
BC137846.1 - 3 - 4 552 99

View Full Product Details