The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145216.3 | NP_660251.1 | 1 - 2 | 402 | 58 |
GenBank mRNA | AF498251.1 | - | 1 - 2 | 224 | 58 |
AK044011.1 | - | 1 - 2 | 403 | 58 | |
BC107200.1 | - | 2 - 3 | 285 | 58 | |
BC107201.1 | - | 2 - 3 | 283 | 58 |