The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146094.2 | NP_666206.1 | 5 - 6 | 917 | 97 |
GenBank mRNA | AB072976.1 | - | 5 - 6 | 832 | 97 |
AF061504.1 | - | 5 - 6 | 819 | 97 | |
AK033308.1 | - | 3 - 4 | 274 | 97 | |
AK083959.1 | - | 5 - 6 | 779 | 97 | |
AK154367.1 | - | 5 - 6 | 867 | 97 | |
AK193280.1 | - | 1 - 2 | 66 | 97 | |
AK198806.1 | - | 1 - 2 | 66 | 97 | |
BC022139.1 | - | 5 - 6 | 862 | 97 | |
BC026831.1 | - | 5 - 6 | 884 | 97 | |
BC026848.1 | - | 5 - 6 | 857 | 97 | |
BC063053.1 | - | 5 - 6 | 777 | 97 | |
KF987079.1 | - | 5 - 6 | 801 | 97 |