The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025819.4 | NP_080095.3 | 6 - 7 | 1150 | 106 |
GenBank mRNA | AK004803.2 | - | 6 - 7 | 1097 | 106 |
BC055064.1 | - | 6 - 7 | 1151 | 106 | |
BC060204.1 | - | 6 - 7 | 1090 | 106 |