The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025931.2 | NP_080207.1 | 3 - 4 | 484 | 63 |
GenBank mRNA | AK011196.1 | - | 3 - 4 | 482 | 63 |
BC017514.1 | - | 3 - 4 | 484 | 63 |