Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bfar
Entrez Gene ID
67118
Gene Name
bifunctional apoptosis regulator
Gene Aliases
3010001A07Rik, AI666707, AW107665, Bar, Rnf47
Chromosome Location
Chr.16: 13671858 - 13703612 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177552.1 NP_001171023.1 4 - 5 873 74
NM_025976.5 NP_080252.1 6 - 7 1248 74
GenBank mRNA AK013874.1 - 6 - 7 1113 74
AK028592.1 - 4 - 5 712 74
AK041839.1 - 5 - 6 987 74
BC027221.1 - 6 - 7 1089 74

View Full Product Details