The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113554.1 | NP_001107026.1 | 9 - 10 | 1977 | 77 |
NM_026149.4 | NP_080425.3 | 9 - 10 | 1980 | 77 | |
GenBank mRNA | AF521132.1 | - | 9 - 10 | 1978 | 77 |
AK014996.2 | - | 9 - 10 | 1980 | 77 | |
AK077673.1 | - | 9 - 10 | 1562 | 77 | |
AK167855.1 | - | 9 - 10 | 1512 | 77 | |
AK168122.1 | - | 9 - 10 | 1512 | 77 | |
BC031583.1 | - | 9 - 10 | 1546 | 77 |