The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026933.2 | NP_081209.1 | 1 - 2 | 167 | 74 |
GenBank mRNA | AK007514.1 | - | 1 - 2 | 168 | 74 |
BC030325.1 | - | 1 - 2 | 186 | 74 |