Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krtap26-1
Entrez Gene ID
69533
Gene Name
keratin associated protein 26-1
Gene Aliases
2310002B14Rik, AI507462
Chromosome Location
Chr.16: 88646824 - 88647796 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027105.2 NP_081381.1 1 - 1 148 73
GenBank mRNA AK009086.1 - 1 - 1 150 73
BC128283.1 - 1 - 1 140 73
BC139410.1 - 1 - 1 143 73
BC139411.1 - 1 - 1 143 73

View Full Product Details