Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ddx51
Entrez Gene ID
69663
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 51
Gene Aliases
2310061O04Rik, AA537002, C81583, R75505
Chromosome Location
Chr.5: 110653434 - 110660496 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027156.3 NP_081432.2 12 - 13 1732 63
GenBank mRNA AK010015.1 - 3 - 4 336 63
AK141361.1 - 12 - 13 1713 63
AK152599.1 - 12 - 13 1700 63
BC023700.1 - 9 - 10 995 63
BC060646.1 - 12 - 13 1709 63
BC069876.1 - 5 - 6 560 63

View Full Product Details