Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smyd3
Entrez Gene ID
69726
Gene Name
SET and MYND domain containing 3
Gene Aliases
2410008A19Rik, Zmynd1
Chromosome Location
Chr.1: 178951960 - 179518052 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027188.3 NP_081464.1 2 - 3 278 88
GenBank mRNA AK010447.1 - 2 - 3 312 88
AK087728.1 - 2 - 3 289 88
AK163057.1 - 2 - 3 295 88
BC052431.1 - 2 - 3 278 88
BC061485.1 - 2 - 3 261 88

View Full Product Details