The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027480.3 | NP_081756.4 | 15 - 16 | 1454 | 62 |
GenBank mRNA | AK019475.1 | - | 2 - 3 | 120 | 62 |
AK043638.1 | - | 13 - 14 | 2536 | 62 | |
AK173316.1 | - | 13 - 14 | 2420 | 62 | |
BC017633.1 | - | 2 - 3 | 125 | 62 |