Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sik3
Entrez Gene ID
70661
Gene Name
SIK family kinase 3
Gene Aliases
5730525O22Rik, 9030204A07Rik, AI447252, AI846254, AI849445, Qsk, SIK-3, mKIAA0999
Chromosome Location
Chr.9: 46012820 - 46224194 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027498.3 NP_081774.3 3 - 4 492 88
GenBank mRNA AK017789.1 - 2 - 3 242 88
BC063268.1 - 3 - 4 288 88
BC080688.1 - 3 - 4 434 88

View Full Product Details