Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Daw1
Entrez Gene ID
71227
Gene Name
dynein assembly factor with WDR repeat domains 1
Gene Aliases
4930563E19Rik, 4933429D11Rik, Wdr69, b2b1116Clo
Chromosome Location
Chr.1: 83159742 - 83210577 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027725.3 NP_082001.2 4 - 5 351 65
NR_027636.1 4 - 5 331 65
GenBank mRNA AK016201.1 - 4 - 5 325 65
AK016977.1 - 4 - 5 352 65
BC049704.1 - 4 - 5 351 65
BC144994.1 - 4 - 5 328 65
BC145995.1 - 4 - 5 351 65
BU612798.1 - 4 - 5 294 65

View Full Product Details