Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Fam175a
- Entrez Gene ID
-
70681
- Gene Name
- family with sequence similarity 175, member A
- Gene Aliases
-
3830405G04Rik, 5630400M01Rik, AI506069, AL024423, AV118690, Ccdc98
- Chromosome Location
-
Chr.5: 100804802 - 100845216 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK017468.1 |
- |
1 - 1 |
742 |
68 |
View Full Product Details