Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tars2
Entrez Gene ID
71807
Gene Name
threonyl-tRNA synthetase 2, mitochondrial (putative)
Gene Aliases
2610024N01Rik, AI429208, Tarsl1
Chromosome Location
Chr.3: 95739974 - 95754977 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163617.1 NP_001157089.1 13 - 14 1793 72
NM_001163618.1 NP_001157090.1 12 - 13 1628 72
NM_027931.3 NP_082207.2 14 - 15 1871 72
GenBank mRNA AK011540.1 - 14 - 15 1757 72
AK080690.1 - 14 - 15 1796 73
AK088675.1 - 12 - 13 1403 72
AK142685.1 - 14 - 15 1766 72
AK164825.1 - 12 - 13 1269 72
AK168075.1 - 12 - 13 1522 72
BC008995.1 - 13 - 14 1678 72
BC067392.1 - 8 - 9 2979 72
BC147501.1 - 14 - 15 1755 72
BG277177.1 - 2 - 3 238 72

View Full Product Details