The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027997.2 | NP_082273.1 | 4 - 5 | 1123 | 81 |
GenBank mRNA | AK009343.1 | - | 4 - 5 | 1123 | 81 |
BC132287.1 | - | 4 - 5 | 1110 | 81 | |
BC144769.1 | - | 4 - 5 | 1111 | 81 |