The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027998.4 | NP_082274.1 | 1 - 1 | 930 | 85 |
GenBank mRNA | AK009330.1 | - | 1 - 1 | 851 | 85 |
AK037108.1 | - | 1 - 1 | 885 | 85 | |
AK145246.1 | - | 1 - 1 | 934 | 85 | |
BC019534.1 | - | 1 - 1 | 915 | 85 | |
BC085262.1 | - | 1 - 1 | 915 | 85 |