Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Chaf1b
Entrez Gene ID
110749
Gene Name
chromatin assembly factor 1, subunit B (p60)
Gene Aliases
2600017H24Rik, C76145, CAF-I p60, CAF-Ip60, CAF1, CAF1A, CAF1P60, MPHOSPH7
Chromosome Location
Chr.16: 93883895 - 93906115 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028083.4 NP_082359.1 9 - 10 933 100
GenBank mRNA AK011243.1 - 9 - 10 934 100
BC013532.1 - 8 - 9 843 100

View Full Product Details