The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028083.4 | NP_082359.1 | 12 - 13 | 1624 | 96 |
GenBank mRNA | AK011243.1 | - | 12 - 13 | 1625 | 96 |
BC013532.1 | - | 11 - 12 | 1534 | 96 |