Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rmi1
Entrez Gene ID
74386
Gene Name
RMI1, RecQ mediated genome instability 1, homolog (S. cerevisiae)
Gene Aliases
4932432N11Rik, C79893
Chromosome Location
Chr.13: 58402248 - 58411149 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168248.1 NP_001161720.1 3 - 4 362 116
NM_028904.3 NP_083180.3 2 - 3 277 116
NR_031761.1 2 - 3 354 116
GenBank mRNA AK016542.1 - 2 - 3 273 116
AK032229.1 - 2 - 3 158 116
AK039934.1 - 1 - 2 2020 116
AK079457.1 - 2 - 3 342 116
AK139703.1 - 2 - 3 309 116
BC023118.1 - 1 - 2 357 116
BC037694.1 - 3 - 4 258 116

View Full Product Details