The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028937.3 | NP_083213.2 | 9 - 10 | 1021 | 68 |
GenBank mRNA | AK016704.1 | - | 9 - 10 | 1023 | 68 |
BC120917.1 | - | 9 - 10 | 1016 | 68 | |
BC120918.2 | - | 9 - 10 | 1016 | 68 | |
DQ086115.1 | - | 9 - 10 | 1021 | 68 |