Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Usp50
Entrez Gene ID
75083
Gene Name
ubiquitin specific peptidase 50
Gene Aliases
1700086G18Rik, 4930511O11Rik
Chromosome Location
Chr.2: 126761050 - 126783460 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029163.3 NP_083439.2 6 - 7 1107 103
GenBank mRNA AK015766.1 - 6 - 7 1092 103
BC061020.1 - 6 - 7 1084 103

View Full Product Details