The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029619.2 | NP_083895.1 | 2 - 3 | 243 | 81 |
GenBank mRNA | AK009912.1 | - | 2 - 3 | 234 | 81 |
BC021619.1 | - | 2 - 3 | 243 | 81 |