Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Efcab6
Entrez Gene ID
77627
Gene Name
EF-hand calcium binding domain 6
Gene Aliases
4931407K02Rik, 4932408N08Rik, bM150J22.2, mKIAA1672
Chromosome Location
Chr.15: 83866712 - 84075025 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161628.1 NP_001155100.1 10 - 11 1714 58
NM_001161629.1 NP_001155101.1 8 - 9 1202 58
NM_029946.4 NP_084222.4 27 - 28 3821 58
GenBank mRNA AK016516.1 - 8 - 9 1307 58
AK019850.1 - 10 - 11 1714 58
AK163363.1 - 5 - 6 892 58
AK220412.1 - 10 - 11 1633 58
BC056955.1 - 15 - 16 2224 58
BC065111.1 - 27 - 28 3821 58

View Full Product Details