The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029948.2 | NP_084224.1 | 2 - 3 | 430 | 64 |
GenBank mRNA | AK019795.1 | - | 2 - 3 | 431 | 64 |
BC066168.1 | - | 2 - 3 | 385 | 64 |