The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170710.1 | NP_001164181.1 | 3 - 4 | 372 | 98 |
NM_001170711.1 | NP_001164182.1 | 3 - 4 | 378 | 98 | |
NM_030121.4 | NP_084397.3 | 3 - 4 | 368 | 98 | |
GenBank mRNA | AF398969.1 | - | 3 - 4 | 295 | 98 |
AK021239.1 | - | 3 - 4 | 309 | 98 | |
AK030168.1 | - | 3 - 4 | 311 | 98 | |
AK087299.1 | - | 3 - 4 | 330 | 98 | |
AK182854.1 | - | 2 - 3 | 128 | 98 | |
AK214224.1 | - | 2 - 3 | 128 | 98 | |
BC025106.1 | - | 3 - 4 | 292 | 98 |