The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168281.1 | NP_001161753.1 | 3 - 4 | 775 | 71 |
NM_133784.3 | NP_598545.2 | 3 - 4 | 854 | 71 | |
GenBank mRNA | AJ299430.1 | - | 3 - 4 | 789 | 71 |
AK142299.1 | - | 3 - 4 | 776 | 71 | |
AK142720.1 | - | 3 - 4 | 779 | 71 | |
BC004640.1 | - | 3 - 4 | 775 | 71 | |
BC014727.1 | - | 3 - 4 | 806 | 71 |