The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001197082.1 | NP_001184011.1 | 4 - 5 | 411 | 64 |
NM_133878.3 | NP_598639.1 | 3 - 4 | 372 | 64 | |
GenBank mRNA | AK076089.1 | - | 3 - 4 | 373 | 64 |
AK150153.1 | - | 3 - 4 | 324 | 64 | |
AK150610.1 | - | 3 - 4 | 322 | 64 | |
AK153193.1 | - | 3 - 4 | 322 | 64 | |
BC019807.1 | - | 3 - 4 | 284 | 64 | |
BC057645.1 | - | 4 - 5 | 366 | 64 |