The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164566.1 | NP_001158038.1 | 5 - 6 | 582 | 82 |
NM_144882.4 | NP_659131.2 | 7 - 8 | 768 | 82 | |
GenBank mRNA | AK031266.1 | - | 7 - 8 | 723 | 82 |
AK032169.1 | - | 7 - 8 | 703 | 82 | |
BC014764.1 | - | 5 - 6 | 582 | 82 | |
BC021879.1 | - | 7 - 8 | 704 | 82 | |
BY739337.1 | - | 5 - 6 | 544 | 81 | |
CN698528.1 | - | 1 - 2 | 119 | 82 |