Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prkag2
Entrez Gene ID
108099
Gene Name
protein kinase, AMP-activated, gamma 2 non-catalytic subunit
Gene Aliases
2410051C13Rik, AAKG, AAKG2, AI854673, H91620p, WPWS
Chromosome Location
Chr.5: 24862735 - 25101906 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145401.2 NP_663376.2 1 - 2 633 63
GenBank mRNA AK154296.1 - 1 - 2 634 63
BC015283.1 - 1 - 2 421 63

View Full Product Details