The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290802.1 | NP_001277731.1 | 5 - 6 | 501 | 90 |
NM_001290804.1 | NP_001277733.1 | 4 - 5 | 426 | 90 | |
NM_007645.4 | NP_031671.1 | 4 - 5 | 450 | 90 | |
GenBank mRNA | AK154250.1 | - | 4 - 5 | 489 | 90 |
AK154466.1 | - | 5 - 6 | 471 | 90 | |
AK155663.1 | - | 4 - 5 | 427 | 90 | |
AK155966.1 | - | 4 - 5 | 415 | 90 | |
BC019402.1 | - | 4 - 5 | 90 | ||
BE849269.1 | - | 4 - 5 | 368 | 90 | |
CT010217.1 | - | 4 - 5 | 339 | 90 | |
CX226822.1 | - | 5 - 6 | 89 |