The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007901.5 | NP_031927.2 | 1 - 2 | 347 | 133 |
GenBank mRNA | AK004591.2 | - | 1 - 2 | 340 | 133 |
AK138316.1 | - | 1 - 2 | 233 | 133 | |
AK146501.1 | - | 1 - 2 | 202 | 133 | |
BC049094.1 | - | 1 - 2 | 222 | 133 | |
BC051023.1 | - | 1 - 2 | 68 | 133 |