The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271402.1 | NP_001258331.1 | 18 - 19 | 1757 | 130 |
NM_001271403.1 | NP_001258332.1 | 17 - 18 | 1602 | 130 | |
NM_001271421.1 | NP_001258350.1 | 17 - 18 | 1652 | 130 | |
NM_007940.4 | NP_031966.2 | 18 - 19 | 1677 | 130 | |
GenBank mRNA | AK002415.1 | - | 18 - 19 | 1727 | 130 |
AK142714.1 | - | 18 - 19 | 1650 | 130 | |
AY098585.1 | - | 17 - 18 | 1616 | 130 | |
BC015087.1 | - | 18 - 19 | 1653 | 130 | |
EF597241.1 | - | 17 - 18 | 1652 | 130 | |
L05781.1 | - | 18 - 19 | 1590 | 130 | |
Z37107.1 | - | 18 - 19 | 1607 | 130 |