The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009746.2 | NP_033876.1 | 2 - 3 | 409 | 74 |
GenBank mRNA | AK131935.1 | - | 2 - 3 | 298 | 74 |
BC005673.1 | - | 2 - 3 | 409 | 74 | |
BC058530.1 | - | 2 - 3 | 280 | 74 | |
Y11905.1 | - | 3 - 4 | 227 | 74 |