The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010225.2 | NP_034355.2 | 1 - 2 | 1442 | 71 |
GenBank mRNA | AK132385.1 | - | 1 - 2 | 1443 | 71 |
BC137947.1 | - | 1 - 2 | 1354 | 71 | |
BC137949.1 | - | 1 - 2 | 1354 | 71 | |
Y11148.1 | - | 1 - 2 | 1149 | 71 |