The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163397.1 | NP_001156869.1 | 1 - 2 | 109 | 58 |
GenBank mRNA | AF041341.1 | - | 1 - 2 | 121 | 58 |
CF726962.1 | - | 1 - 2 | 109 | 58 |