The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011804.2 | NP_035934.1 | 3 - 4 | 663 | 78 |
GenBank mRNA | AF084524.1 | - | 3 - 4 | 663 | 78 |
AK005388.1 | - | 3 - 4 | 684 | 78 | |
AK010947.1 | - | 3 - 4 | 680 | 78 | |
AK085844.1 | - | 3 - 4 | 475 | 78 | |
AK144101.1 | - | 3 - 4 | 682 | 78 | |
AK150006.1 | - | 4 - 5 | 789 | 78 | |
BC027426.1 | - | 3 - 4 | 663 | 78 |