Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Crym
Entrez Gene ID
12971
Gene Name
crystallin, mu
Gene Aliases
-
Chromosome Location
Chr.7: 120186384 - 120201988 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016669.1 NP_057878.1 1 - 2 236 109
GenBank mRNA AF039391.1 - 1 - 2 236 109
AK138422.1 - 1 - 2 207 109
AK143116.1 - 1 - 2 226 109
AK163781.1 - 1 - 2 207 109
AK163801.1 - 1 - 2 233 109
BC045159.1 - 1 - 2 197 109

View Full Product Details