The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016706.2 | NP_057915.2 | 5 - 6 | 1568 | 72 |
GenBank mRNA | AA240407.1 | - | 3 - 4 | 168 | 72 |
AF208663.1 | - | 5 - 6 | 1550 | 72 | |
BC031167.1 | - | 5 - 6 | 1583 | 72 | |
CF105200.1 | - | 4 - 5 | 366 | 72 |