The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018769.3 | NP_061239.1 | 5 - 6 | 755 | 71 |
GenBank mRNA | AF073309.1 | - | 5 - 6 | 756 | 71 |
AK016561.1 | - | - | 1123 | 71 | |
AK089459.1 | - | - | 743 | 71 | |
AK155135.1 | - | - | 734 | 71 | |
AK171278.1 | - | 5 - 6 | 754 | 71 | |
BC051962.1 | - | 5 - 6 | 815 | 71 | |
BC086618.1 | - | 5 - 6 | 815 | 71 | |
BC132303.1 | - | 4 - 5 | 726 | 71 |